Five-Star™ Cutters, Medium Head, Excelta
Carbon steel, 0.008 - 0.042" soft, 5", Medium Round Head, ESD safe molded grips, Semi-flush
63042-834
Supplier #: 7240E
Five-Star™ 7000 series medium head cutters are precision side cutters designed to make an extremely precise cut on small leads and soft wire.
- Available either with long foam cushion grips or ESD safe molded grips
- The dual stainless-steel leaf springs provide smooth action
These are feature with raised bearing surface, Rockwell C 63-65 hardness, and a Delrin washer for smooth operation. The CNC machining allows the joint surfaces to be perfect to allow for long life.
Documents
- Material:Carbon steel
- Cutting Capacity:0.008 - 0.042" soft
- Tip Style:Medium Round Head
- Overall Length:5"
- Grip Type:ESD safe molded grips
- Cut Type:Semi-flush