Scissors, Fine, Excelta Corp®
Scissors, long blade, 2 star quality, 5" (125 mm), Stainless steel AISI 440, 50.8 mm (2"), Straight
63041-844
: 298B
These high quality scissors are designed for delicate work and close tolerance cutting.
- Cleanroom safe, stainless steel
- Variety of blade lengths
These stainless steel scissors offer fine blades for clean cutting. They are all made of AISI 440 stainless steel. They are offered in a variety of blade lengths.
- Description:Scissors, long blade, 2 star quality
- Length:5" (125 mm)
- Material:Stainless steel AISI 440
- Curvature:Straight
- Blade Length:50.8 mm (2")