Tweezers with Straight Fine Tip, Style 3, Excelta
Proveedor: EXCELTA CORPORATION
Certificados
Acerca de este artículo
Style 3 is very popular in micro-electronic assembly. It features a straight tweezer with a very fine point.
- Ideal for working under magnification
- Available in three different materials
- Available in three different qualtiy grades
- Ergonomic handles are standard option
- Teflon coated tip option for non-marring
The 'Style 3' series straight very fine precision pointed tweezer is designed for work under magnification. The overall length is 4.75". These are also availabe with ergonomic handles (ET). We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).
Frequently Bought Together