Protector® Premier® Laboratory Hoods, Labconco®
About this item
Labconco Protector® Premier® Fiberglass Fume Hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up. There are no seams where deterioration can occur or where contaminants can collect. These general purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.
Hoods are available with a built-in standard fiberglass blower, or an explosion-proof fiberglass blower, or without a blower (remote blower sold separately).
Hoods are available with a built-in standard fiberglass blower, or an explosion-proof fiberglass blower, or without a blower (remote blower sold separately).
There are no seams where deterioration can occur or where contaminants can collect. These general purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.
Certifications: All models are ETL listed. 230V models are CE listed.
Ordering information: Customize your hood with optional service fixtures that accommodate air, gas, vacuum or water lines. Complete your fume hood with a variety of accessories including blowers, work surfaces, storage cabinets, fire extinguishers, and air flow monitors (sold separately).
Services: VWRCATALYST™ offers customer on-site validation, testing and certification services including face velocity profile test, smoke generation test, tracer gas test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWR[email protected] for more details. You can also visit https://vwr.com/equip_services.
1 Options Available Below
Product Details & Documents
Labconco Protector® Premier® Fiberglass Fume Hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up. There are no seams where deterioration can occur or where contaminants can collect. These general purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.
Hoods are available with a built-in standard fiberglass blower, or an explosion-proof fiberglass blower, or without a blower (remote blower sold separately).
Hoods are available with a built-in standard fiberglass blower, or an explosion-proof fiberglass blower, or without a blower (remote blower sold separately).
There are no seams where deterioration can occur or where contaminants can collect. These general purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.
Certifications: All models are ETL listed. 230V models are CE listed.
Ordering information: Customize your hood with optional service fixtures that accommodate air, gas, vacuum or water lines. Complete your fume hood with a variety of accessories including blowers, work surfaces, storage cabinets, fire extinguishers, and air flow monitors (sold separately).
Services: VWRCATALYST™ offers customer on-site validation, testing and certification services including face velocity profile test, smoke generation test, tracer gas test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWR[email protected] for more details. You can also visit https://vwr.com/equip_services.
Specifications for Product Family
Specifications
- Catalog No:
- 89232-732
- 89232-740
- 89232-748
- 89232-758
- 89232-784
- Environmentally Preferable:
- Sustainable Materials|Energy Efficient
- Sustainable Materials|Energy Efficient
- Sustainable Materials|Energy Efficient
- Sustainable Materials|Energy Efficient
- Sustainable Materials|Energy Efficient
Specifications
Product Family Options
Product Information
- ElectricalNominal WidthElectrical Duplex OutletService FixturesAvailabilityPrice
- 89232-732Protector® Premier® Laboratory Hood without BlowerProtector® Premier® Laboratory Hood without Blower115 V4'12Log in for availabilityEach (1UN)$244,530.00Supplier Number: 100400002Specifications:Product available on GSA Advantage®
Sustainable Materials|Energy Efficient More
Specifications
Cat. No.89232-732Environmentally PreferableSustainable Materials|Energy EfficientProduct available on GSA Advantage®Sustainable Materials|Energy Efficient - 89232-740Protector® Premier® Laboratory Hood with BlowerProtector® Premier® Laboratory Hood with Blower115 V4'12Discontinued-Supplier Number: 100400042Specifications:Product available on GSA Advantage®
Sustainable Materials|Energy Efficient More
Specifications
Cat. No.89232-740Environmentally PreferableSustainable Materials|Energy EfficientProduct available on GSA Advantage®Sustainable Materials|Energy Efficient - 89232-748Protector® Premier® Laboratory Hood without BlowerProtector® Premier® Laboratory Hood without Blower115 V5'00Discontinued-Supplier Number: 100500000Specifications:Product available on GSA Advantage®
Sustainable Materials|Energy Efficient More
Specifications
Cat. No.89232-748Environmentally PreferableSustainable Materials|Energy EfficientProduct available on GSA Advantage®Sustainable Materials|Energy Efficient - 89232-758Protector® Premier® Laboratory Hood with BlowerProtector® Premier® Laboratory Hood with Blower115 V5'12Discontinued-Supplier Number: 100500042Specifications:Product available on GSA Advantage®
Sustainable Materials|Energy Efficient More
Specifications
Cat. No.89232-758Environmentally PreferableSustainable Materials|Energy EfficientProduct available on GSA Advantage®Sustainable Materials|Energy Efficient - 89232-784Protector® Premier® Laboratory Hood without BlowerProtector® Premier® Laboratory Hood without Blower115 V8'00Discontinued-Supplier Number: 100800000Specifications:Product available on GSA Advantage®
Sustainable Materials|Energy Efficient More
Specifications
Cat. No.89232-784Environmentally PreferableSustainable Materials|Energy EfficientProduct available on GSA Advantage®Sustainable Materials|Energy Efficient
Accessories
Product Information
- Catalog #AvailabilityPrice
- 4' SpillStopper™ Work Surface, Right Side Cupsink Cutout4' SpillStopper™ Work Surface, Right Side Cupsink Cutout89232-800Log in for availabilityEach (1UN) - $41,340.00
Product Information
- Catalog #AvailabilityPrice
- 4' SpillStopper™ Work Surface, No Cupsink Cutouts4' SpillStopper™ Work Surface, No Cupsink Cutouts89232-792Log in for availabilityEach (1UN) - $36,860.00