Receiver, Moisture Test, Barrett-Type, Ace Glass
Catalog # 89060-344
Supplier: ACE GLASS, INC.
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Capacidad:20 ml
- Descripción:With Unjacketed Side Arm, Glass Plug Style
- Referencia:89060-344
- Nº de proveedor:7745-04
Specifications
About this item
This moisture test receiver is used to determine the water content in petroleum and bituminous material
It is supplied with [ST] 24/40 joints, and with a stopcock to empty the measuring tube during the determination. The 10mL size is subdivided in 0.1mL from 0–1mL, 0.2mL from 1–10mL; 20mL size in 0.1mL from 0–1mL, 0.2mL from 1–20mL. Fabricated from borosilicate glass.
Ordering information: Available with 2mm bore glass, or with 1:5 solid PTFE stopcock plug. Available with jacketed or unjacketed side arm. Jacketed side arm version helps prevent condensation and facilitates improved performance.