Tweezers with Straight Fine Tips, Style 1, Excelta
Catalog # 63041-412
Supplier: EXCELTA CORPORATION
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Descripción:Tweezers, very fine point, 2 star quality
- Matriz:Stainless steel 316L, anti-magnetic
- Curvature:Straight
- Referencia:63041-412
- Estilo de la punta:Straight Fine Tip
- Longitud total:112.5 mm (4.5")
- Nº de proveedor:1-SA-PI
Specifications
About this item
Style 1 is very popular in micro-electronic assembly. It features a straight tweezer with a very fine point.
- Design is excellent for work under scope
- Available in 3 different quality grades
- Available in 3 different materials depending on application
The 'Style 1' Series tweezers is ideal for delicate applications. They feature fine straight points and are designed for bending and handling very fine wires and components. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic). The suffix 'ET' features our Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat.