Tweezers with Curved Fine Tip, Style 7, EXCELTA®
Supplier: EXCELTA CORPORATION
Certificates
About this item
Style 7 tweezers feature a curved tip and precise points making them ideal for working under light magnification. The 7B version has finally serrated tips for better gripping.
- Tapered curved tips for greater visibility/accessibility
- Very fine points for precision
- Offered in 3 different materials
- Three quality levels
- Available with ergonomic grips
The 'Style 7' Series curved tapered very fine precision point tweezer allows maximum visibility under magnified applications. The 'Style 7B' Series additional offers serrated tips for gripping power. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic). The 'ET' version features our Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat.
Frequently Bought Together