Tweezers with Flat Duck-Bill Tip, Style 2A, Excelta
Catalog # 63041-874
Supplier: EXCELTA CORPORATION
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Descripción:Tweezers, tapered flat point, 3 star quality
- Matriz:Stainless steel 316L, anti-magnetic
- Curvature:Straight
- Referencia:63041-874
- Estilo de la punta:Straight Duck-Bill Tip
- Longitud total:118.75 mm (4.75")
- Nº de proveedor:2A-SA
- Tip Width:0.059" (1.5 mm)
- Tip Length:0.2 mm (0.008")
Specifications
About this item
The Style 2A tweezers feature a flat round tip. They are excellent for pulling labels without scratching and also for handling surface mount components.
- Excellent for handling labels/smd components
- Available in 3 different materials
- Three different quality grades
- Available with ergonomic handles
- Teflon coated tips available, soft handling
The 'Style 2A' Series tapered flat round tipped tweezer is designed for handling labels or SMD components. The different options offered are ergonomic handles or teflon coated tips (to prevent scratching).depending on your application. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).