Carbofib Three Star Tweezers, Soft ESD Replaceable Tip, Excelta
16003-848
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Descripción:Tweezers, replaceable tip, 3 star quality
- Matriz:Carbon fiber tips
- Curvature:Straight
- Referencia:16003-848
- Longitud total:125 mm (5")
- Nº de proveedor:159D-RT
- Tip Width:0.06" (1.5 mm)
- Tip Length:0.38 mm (0.015")
These replaceable tip tweezers utilize a carbon-fiber material that makes them ESD Safe. They are used for handling delicate parts.
- Replaceable tips
- Available in a multitude of tip sizes
- Temperature resistant to 249 °F
- Conductive
- Will not scratch or mar parts
- Excellent tensile and flexural strength
These soft replaceable tip tweezer boasts Carbofib™ tips. The Carbofib™ tips are a polymer based material with 30% carbon fibers added (conductive 10² ohms/sq). Carbofib™ has very high rigidity, fatigue and creep resistance. The bodies are made of anti-magnetic Neverust® stainless steel (AISI 316L). They are excellent for handling delicate items.