SP Bel-Art Drum and Carboy Funnels, Bel-Art Products, a part of SP
Catalog # 47750-836
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacidad:2.1 L (0.56 gal.)
- Descripción:Drum funnel stem dia. 21 mm (0.83")
- Altura:250 mm (9.85")
- Referencia:47750-836
- D.E. superior:207 mm (8.15")
- Longitud del vástago:88 mm (3.45")
- Nº de proveedor:147120200
Specifications
About this item
Large funnels are ideal for filling drums, barrels, tanks and carboys providing ample volume and fast flow.
- Natural polypropylene construction
- Autoclavable
- Large funnel provides ample volume and fast flow
- Handle in some models adds stability
The two smaller sizes have a handle at the rim for added stability. Molded of natural polypropylene, funnels are steam autoclavable at 121ºC (250ºF).