Jar with Cap, HDPE
Supplier: DYNALON
Jump to:
Acerca de este artículo
These cylindrical food-grade HDPE containers with black HDPE screw closures are supplied with a neck insert for added security, suitable for powders, solid specimens, creamy or semisolid substances, crystals, and hygroscopic substances.
- Aligned with GLOBE protocols
5 Options Available Below
Product Details & Documents
These cylindrical food-grade HDPE containers with black HDPE screw closures are supplied with a neck insert for added security, suitable for powders, solid specimens, creamy or semisolid substances, crystals, and hygroscopic substances.
- Aligned with GLOBE protocols
Product Family Options
Información del producto
- AvailabilityPrice
- 470191-503Jar with Cap, HDPE, 70 mLJar with Cap, HDPE, 70 mLEach (1UN)$2.20Case de 10 (1UN)$22.00Supplier Number: 226415-0070
- 470191-496Jar with Cap, HDPE, 120 mLJar with Cap, HDPE, 120 mLEach (1UN)$101.59Paquete de 10 (1UN)$110.73Supplier Number: 226415-0120
- 470137-446Jar with Cap, HDPE, 250 mLSupplier Number: 226415-0250
- 470191-495Jar with Cap, HDPE, 1000 mLSupplier Number: 226415-1000
- 470191-498Jar with Cap, HDPE, 2000 mLJar with Cap, HDPE, 2000 mLEach (1UN)$228.40Paquete de 10 (1UN)$248.95Supplier Number: 226415-2000