NeuLog Light Logger Sensor
1 lx (1 - 1000 lx)
6 lx (0 - 6000 lx)
150 lx (0 - 150000 lx)
Product Details & Documents
This adaptable light sensor has a wide range of uses.
- Three ranges for illumination measurement
- Fast and slow modes available
- Ideal in low-light and high-light environments
It can be used to study photosynthesis in biology, light-emitting chemical reactions in chemistry, the effect of changing voltage on a light-bulb's output in physics, and more. This sensor measures illumination with three ranges.
With both fast and slow modes, it can measure fast light changes, such as those produced by light bulbs connected to an AC supply, and the almost-steady levels outside on a sunny day.
It supports experiments that last from 50 ms to 31 days.
: One or more of the following items are needed but not included–a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand-alone probe or if you have the wireless module), wireless module (not needed if connecting to a computer through the USB module).
: For educational use only.
Especificaciones
- Resolution:12 bit (ADC)
1 lx (1 - 1000 lx)
6 lx (0 - 6000 lx)
150 lx (0 - 150000 lx) - Description:NeuLog light logger sensor
- Range:0 - 1000 lx, 0 - 6000 lx, 0 - 150000 lx
- Cat. No.:470137-376
- Sampling frequency:Signal: 3000 S/s
Illumination: 100 S/s