NeuLog Wide-Range Temperature Logger Sensor
NeuLog wide-range temperature logger sensor, T: −200...1200 °C, T: 0.1 °C
ADC: 10 bit
ADC: 10 bit
Catalog # 470137-434
Supplier: EISCO SCIENTIFIC LLC
: NUL234
Jump to:
Product Details & Documents
Measures a wide range of temperatures.
- Max. sample rate 100 S/sec.
- Experimental duration 1 sec. to 31 days
Based on a thermocouple sensor, this unit enables the measurement of very high and very low temperature levels. It runs in two modes of operation either Celsius or Fahrenheit.
: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
: For educational use only.
Especificaciones
- Resolution:T: 0.1 °C
ADC: 10 bit - Description:NeuLog wide-range temperature logger sensor
- Range:T: −200...1200 °C
- Cat. No.:470137-434
- Interface:USB