NeuLog Respiration Monitor Belt Logger Sensor
Catalog # 470140-290
Supplier: EISCO SCIENTIFIC LLC
Specifications
- Resolución:1; ADC: 10 bit
- Descripción:NeuLog respiration monitor belt logger sensor
- Referencia:470140-290
- Rango:0 - 6500 arb
- Nº de proveedor:NUL-236
Specifications
About this item
Respiration impacts heart rate.
- Max. sample rate 100 S/sec
- Experiment duration 1 sec. to 31 days
- Belt included
This sensor monitors abdominal or thoracical breathing as well as giving an indication of the depth of breathing.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.