Distillation Apparatus
Distillation Apparatus
Catalog # 470136-974
Supplier: UNITED SCIENTIFIC SUPPLIES
: DSA001
Jump to:
Product Details & Documents
Quick setup for distillation
- Sturdy support stand holds all the required pieces
- Can be easily adjusted to fit your needs
This complete kit includes all of the materials necessary to construct an inexpensive, versatile distillation apparatus.
Includes:
500 ml round bottom borosilicate flask with side tube
300 mm Graham condenser
250 ml Erlenmeyer flask
Glass stopper
Support stand (6×9") with 24" rod
Swivel burette clamp
Extension clamp
Clamp holder
Activity guide
Especificaciones
- Description:Distillation Apparatus