ContactUsLinkComponent
Green Nanochemistry: Synthesis of Silver Nanoparticles
Green Nanochemistry: Synthesis of Silver Nanoparticles
Catalog # 470136-726
Supplier:  ALDON CORP SE
undefined
Green Nanochemistry: Synthesis of Silver Nanoparticles
Catalog # 470136-726
Supplier:  ALDON CORP SE
Supplier Number:  IS9506
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Descripción:
    Green Nanochemistry: Synthesis of Silver Nanoparticles
  • No. of Lab Groups:
    15
  • Referencia:
    470136-726
  • Nº de proveedor:
    IS9506
Specifications
About this item
Synthesize Silver Nanoparticles from Silver Nitrate.
  • Green chemistry inquiry activity
  • Non-hazardous alternative chemicals
  • All chemicals required are included
Certain materials, when prepared at nanoscale levels, display different physical and chemical properties than those of the same materials prepared on a larger scale. In this activity, students synthesize silver nanoparticles from silver nitrate. The particles are synthesized in glucose and starch, as opposed to the more hazardous reagents typically used.
Includes:
Silver Nitrate
Dextrose
Soluble Starch
Sodium Hydroxide
Teacher's Guide and Student Copymasters