ContactUsLinkComponent
Visual Scientifics Base
Visual Scientifics Base
Catálogo # 470148-822
undefined
Visual Scientifics Base
Catálogo # 470148-822
Número de proveedor:  PTBASE
Especificaciones
  • Descripción:
    Visual scientifics base
  • Tamaño:
    25×16×2.5"
  • Referencia:
    470148-822
  • Nº de proveedor:
    PTBASE
Especificaciones
Acerca de este artículo
This base platform supports the full range of Visual Scientifics activities and accessories.
  • High-contrast grid
  • Magnetic
  • Platform neutral - works with your preferred probeware/video analysis tools
Your students can set up their experiments quickly and easily with this large, sturdy, magnetic base that features threaded junction points for precise probeware mounting. Used in conjunction with Visual Scientifics accessory equipment, this laboratory investigation system offers students an engaging platform and accurate results. Its high-contrast color grid also allows for easy data collection with video analysis systems.