Tie Dye Lab Coat
Acerca de este artículo
Protect Clothing from Spills and Splashes
- Knee-length
- Easy-care cotton blend
- Tie-dye pattern
Similar to our top-selling white lab coats, these knee-length tie dye lab coats are made of an easy care white cotton-blend fabric of 65% polyester, 35% cotton. Featuring a vibrant tie-dye pattern, the coats have one upper left breast pocket, two roomy lower pockets, and side slit openings.
4 Options Available Below
Product Details & Documents
Protect Clothing from Spills and Splashes
- Knee-length
- Easy-care cotton blend
- Tie-dye pattern
Similar to our top-selling white lab coats, these knee-length tie dye lab coats are made of an easy care white cotton-blend fabric of 65% polyester, 35% cotton. Featuring a vibrant tie-dye pattern, the coats have one upper left breast pocket, two roomy lower pockets, and side slit openings.
Specifications for Product Family
Especificaciones
- Catalog No:
- 470136-710
- 470136-712
- 470136-714
- 470136-704
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 65% Polyester, 35% Cotton
- 65% Polyester, 35% Cotton
- 65% Polyester, 35% Cotton
- 65% Polyester, 35% Cotton
Especificaciones
Product Family Options
Información del producto
- TypeClothing SizeGarment fitAvailabilityPrice
- Supplier Number: LBCTSMEspecificaciones:
More
Specifications
Cat. No.470136-710Garment applicationLaboratoryGenderUnisexSleeve or leg lengthLongClosure typeButtonsFabric65% Polyester, 35% Cotton - Supplier Number: LBCTMDEspecificaciones:
More
Specifications
Cat. No.470136-712Garment applicationLaboratoryGenderUnisexSleeve or leg lengthLongClosure typeButtonsFabric65% Polyester, 35% Cotton - Supplier Number: LBCTLGEspecificaciones:
More
Specifications
Cat. No.470136-714Garment applicationLaboratoryGenderUnisexSleeve or leg lengthLongClosure typeButtonsFabric65% Polyester, 35% Cotton - Supplier Number: LBCTXLEspecificaciones:
More
Specifications
Cat. No.470136-704Garment applicationLaboratoryGenderUnisexSleeve or leg lengthLongClosure typeButtonsFabric65% Polyester, 35% Cotton