Ultraform® Powder-Free Nitrile Exam Gloves, Microflex®
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificates
About this item
Ultraform® powder-free nitrile exam gloves have textured fingertips and its unique soft formulation forms to your hand effortlessly as if it is second skin.
- Textured fingertips
- Unique soft formulation forms to your hand
In field tests users wearing Ultraform® actually experienced an increase in fine motor task performance compared to a bare hand.
Packaging: Packaged 3000 gloves per case, 300 gloves per dispenser box by weight (*250 XL).
Specifications
- Catalog No:
- 89235-582
- 89235-580
- 89235-578
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Espesor del puño:
- 2 mil
- 2 mil
- 2 mil
- Espesor del dedo:
- 3.2 mil
- 3.2 mil
- 3.2 mil
- Espesor de la palma:
- 2 mil
- 2 mil
- 2 mil
Specifications
Related Information
Frequently Bought Together