Precision Seal™ Septa, Rubber, Ace Glass Incorporated
Catalog # 89084-554
Supplier: ACE GLASS, INC.
Small Business Enterprise
Specifications
- Color:White
- Referencia:89084-554
- Para ser utilizado con:24/40 [ST] Joint
- Nº de proveedor:9106-135
Specifications
About this item
Seals are engineered for a precision fit (80% glass to rubber contact) in standard taper glassware joints and tubes
Precision-Seal™ septa are manufactured under “white room” conditions, from one certified raw material formulation, for absolute consistency in all sizes, lot-to-lot.
Ordering information: Available in red or white. Mixed set includes samples of each size available.