Orders LinkContactUsLinkComponent
Visual Scientifics Hooke's Law
Visual Scientifics Hooke's Law
Visual scientifics Hooke's law
Catalog # 470135-562
Número de proveedor #:  PTHOKL
Visual Scientifics Hooke's Law
Catalog # 470135-562
Visual Scientifics Hooke's Law
Visual Scientifics Hooke's Law
Visual scientifics Hooke's law
Price
$845.88
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Students investigate and learn the principles of Hooke’s Law with this activity.
  • Lab manual included, with activities and extensions
  • Covers NGSS and common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
Using technology to investigate Hooke's Law, students will discover the linear relationship between applied mass and spring extension. The included lab manual provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and Common Core concepts.
It's especially applicable to mechanics and physics classes.
 : Visual Scientifics Base (470148-822) required, but sold separately.
 : This kit includes 3 springs with different spring constants, aluminum support rod with hook, metal ruler, hanging mass hook with pointer, and 9 to 10 g masses. Recommended accessories: video camera and graphical analysis software.
Especificaciones
  • Description:
    Visual scientifics Hooke's law
Accessories
Información del producto
  • Catalog #
    Size
    Availability
    Price
  • Visual scientifics base
    Visual scientifics base
    470148-822
    25×16×2.5"
    Log in Para disponibilidad
    Each (1UN) - $139.50