Orders LinkContactUsLinkComponent
Three-Prong Extension Clamp
Three-Prong Extension Clamp
Three-Prong Extension Clamp
Certificates
Three-Prong Extension Clamp
Three-Prong Extension Clamp
2 Options Available Below
Product Details & Documents
Much Stronger Than Plastic Clamps
  • Silicone coated Jaws
  • Opens up to 3"
This handy three-prong extension clamp is highly useful for attaching items needed for experiments to bases and rods. Each is constructed of die-cast zinc alloy, and chrome plated with jaws that are coated silicone. Opens to approximately 3", with a recommended maximum grip size of 2 1/4". The clamps also feature double-keyed, spring-type adjustment screws. The Boss head clamp is lockable up to 360°. The recommended working temperature is up to 280°C. Size of extension rod: 8 1/4" L x 5/16" OD; overall length: 12 3/4".
Product Family Options
Product Information
  • Availability
    Price
  • 470135-120
    Three-Prong Extension Clamp
    Three-Prong Extension Clamp
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $607.61
    $607.61
    Each (1UN)
       $607.61
    Supplier Number: ECSIL1
  • 470135-122
    Three-Prong Extension Clamp, with Boss Head
    Three-Prong Extension Clamp, with Boss Head
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $734.49
    $734.49
    Each (1UN)
       $734.49
    Supplier Number: ECSIL2