Orders LinkContactUsLinkComponent
Bottle Top Dispenser
Bottle Top Dispenser
Supplier:   VWR International
Bottle Top Dispenser
Supplier:  VWR International
Certificates
Bottle Top Dispenser
Bottle Top Dispenser
2 Options Available Below
Product Details & Documents
Spill-Free and Safe Dispensing
Prevent spills while making your laboratory a cleaner and safer environment. This innovative bottle top dispensing system allows for use with a wide range of laboratory chemicals. The unique nozzle easily dispenses liquids accurately into the receiving container and is autoclavable to 121˚ C. The system includes a variety of bottle adapters to fit standard-size laboratory bottles. Graduated reagent bottles are recommended for replacements.
Product Family Options
Información del producto
  • Availability
    Price
  • 470134-946
    Bottle top dispenser, 1-10 ml
    Bottle top dispenser, 1-10 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,197.98
    $4,197.98
    Each (1UN)
       $4,197.98
  • 470134-948
    Bottle top dispenser, 2.5-30 ml
    Bottle top dispenser, 2.5-30 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $5,812.99
    $5,812.99
    Each (1UN)
       $5,812.99