ESK® Sampling Kit, with Tube, Swab and Buffered Peptone Water, Puritan Medical Products
Catálogo # 89221-724
Proveedor: PURITAN MEDICAL PRODUCTS COMP
Product available on GSA Advantage®
Especificaciones
- Esterilidad:Sterile
- Referencia:89221-724
- Tamaño de la punta:17.4 mm (0.687")
- Longitud del mango:82.55 mm (3.25")
- Tip Diameter:5.15 mm (0.203")
- Longitud total:103.12 mm (4.06") (tube)
- Fill Solution:10 mL Buffered Peptone Water
- Nº de proveedor:25-83010PDBBPW
Especificaciones
Acerca de este artículo
Easy-to-use, reliable test kits are available with Neutralizing Buffer, Buffered Peptone Water, Letheen Broth, or Butterfields Solution, in a 4mL or 10mL fill
These kits are used for the pre-enrichment of injured Salmonella species from food specimens prior to inoculation to a selective enriched medium for isolation and identificaiton.
Pre-filled with 4mL or 10mL Buffered Peptone Water. Shelf life is 18 months. Made in USA.
They come with a small polyester tip attached to a quick-turn, leak-resistant cap to ensure optimum control.
Información de pedido: Kits are packaged 50 individually wrapped tubes per case. Bulk packaging includes 6 boxes of 50 individually wrapped tubes per box.