ESK® Sampling Kit, with Tube, Swab and Neutralizing Buffer, Puritan Medical Products
Catalog # 89221-716
Supplier: PURITAN MEDICAL PRODUCTS COMP
Product available on GSA Advantage®
Specifications
- Esterilidad:Sterile
- Referencia:89221-716
- Tamaño de la punta:17.4 mm (0.687")
- Longitud del mango:82.55 mm (3.25")
- Tip Diameter:5.15 mm (0.203")
- Longitud total:103.12 mm (4.06") (tube)
- Fill Solution:10 mL Neutralizing Buffer
- Nº de proveedor:25-83010-PDBNB
Specifications
About this item
Easy-to-use, reliable test kits are available with Neutralizing Buffer, Buffered Peptone Water, Letheen Broth, or Butterfields Solution, in a 4mL or 10mL fill
Neutralizing buffer is recommended for use in microbiological examination of surfaces for food and dairy products.
Pre-filled with 4mL or 10mL Neutralizing Buffer. Shelf life is 18 months. Made in USA.
They come with a small polyester tip attached to a quick-turn, leak-resistant cap to ensure optimum control.
Ordering information: Kits are packaged 50 individually wrapped tubes per case. Bulk packaging includes 6 boxes of 50 individually wrapped tubes per box.