ContactUsLinkComponent
Citrad™ Citric Acid Liquid Cleaner
Citrad™ Citric Acid Liquid Cleaner
Catalog # 89234-814
Supplier:  DECON LABS
   Sustainable Packaging
   Waste Reducing
undefined
Citrad™ Citric Acid Liquid Cleaner
Catalog # 89234-814
Supplier:  DECON LABS
Supplier Number:  4401
   Sustainable Packaging
   Waste Reducing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Tamaño:
    3.8 L (1 gal.)
  • Environmentally Preferable:
    Sustainable Packaging,
    Waste Reducing
  • Referencia:
    89234-814
  • Empaquetado:
    Poly bottle
  • Nº de proveedor:
    4401
Specifications
About this item
Citrad™ concentrated acidic detergent is ideal for the removal of salts, scale, and metal-oxide. It can also be used for metal brightening and pharmaceutical process equipment.
  • Completely free-rinsing
  • Biodegradable
Citrad™ concentrated acidic detergent contains a safe, organic acid and is phosphate-free, so it will not interfere with sensitive equipment or testing procedures. This concentrated formulation must be diluted with distilled, deionized, or clean potable water and can be used as a soak, scrub, or ultrasonic cleaner. Use to clean labware, stills, cleanrooms, optical parts, pharmaceutical equipment, and manufacturing equipment. Citrad™ is also effective in removing metal oxides, scale, soil, grease, chemicals, solvents, and other compounds from surfaces and equipment.