ContactUsLinkComponent
Contrad® NF Non-foaming Liquid Detergent
Contrad® NF Non-foaming Liquid Detergent
Catalog # 89233-162
Supplier:  DECON LABS
   Sustainable Packaging
   Waste Reducing
undefined
Contrad® NF Non-foaming Liquid Detergent
Catalog # 89233-162
Supplier:  DECON LABS
Supplier Number:  6001
   Sustainable Packaging
   Waste Reducing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Tamaño:
    10 L (2.64 gal.)
  • Environmentally Preferable:
    Sustainable Packaging,
    Waste Reducing
  • Referencia:
    89233-162
  • Empaquetado:
    Poly bottle
  • Nº de proveedor:
    6001
Specifications
About this item
Contrad® NF is a non-foaming concentrated liquid detergent designed for high-pressure cleaning applications in laboratory and industrial automatic washing machines, CIP systems, steam washers, etc.
  • Phosphate-free and chlorine-free
Contrad® NF is particularly recommended for critical cleaning of labware, where total rinsability and residue-free results are vital. It is also suitable for plant cleaning, such as biotechnology and pharmaceutical production equipment.