ContactUsLinkComponent
Tube Fittings, Connectors, PTFE, Ace Glass Incorporated
  89077-524
Small Business Enterprise
undefined
Tube Fittings, Connectors, PTFE, Ace Glass Incorporated
  89077-524
 :  12770-36
Small Business Enterprise
  • Tipo de conexión:
    Barb
  • Descripción:
    PTFE Connector
  • Dimensiones:
    16 x 39.6L mm
  • Referencia:
    89077-524
  • NPT:
    ¼"
  • Nº de proveedor:
    12770-36
These all-PTFE fittings have a male NPT (MNPT) thread at one end and either tube compression, a hose barb, or MNPT thread at the other end.