ContactUsLinkComponent
Clamp Holder, Nickel-Plated Zinc, Ace Glass Incorporated
Clamp Holder, Nickel-Plated Zinc, Ace Glass Incorporated
Catálogo # 89084-746
Proveedor:  ACE GLASS, INC.
Product available on GSA Advantage®
Small Business Enterprise
undefined
Clamp Holder, Nickel-Plated Zinc, Ace Glass Incorporated
Catálogo # 89084-746
Proveedor:  ACE GLASS, INC.
Número de proveedor:  11080-19
Product available on GSA Advantage®
Small Business Enterprise
Especificaciones
  • Descripción:
    Regular Clamp Holder
  • Referencia:
    89084-746
  • Nº de proveedor:
    11080-19
Especificaciones
Acerca de este artículo
This clamp holder is normally used to secure clamps to rods and lab frame supports
Made of heavily nickel-plated zinc. Will grip rods up to and including 19mm in diameter. Attaches clamp at a right angle.