Specifications
- Porosidad:2.0 µm
- Overall O.D.:0.250" (0.64 cm)
- Frit Volume:1.7 μl
- Referencia:21511-429
- Nº de proveedor:A-100
- Disc Thickness:0.062" (0.16 cm)
- Ring Material:PEEK
Specifications
About this item
Upchurch Scientific® Stainless Steel frits are available in several porosities: 0.5, 2, 10, and 20 µm. Most of these frits include either a PCTFE, ETFE or PEEK polymer sealing ring and many have the common 0.250”(0.64 cm) and 0.254"(0.64 cm) OD's, which allow them to be used in Upchurch Scientific® precolumn and inline filters. Choose frits with larger diameter faces and/or larger porosities for faster flow rates. Smaller diameter faces and/or smaller porosities are ideal for applications sensitive to extra flow path volume.
Upchurch Scientific® Frit-In-A-Ferrule™ seal and filter simultaneously and can filter at any point in your system where 1/16" or 1/8" OD tubing is used in a flat-bottom 1/4-28, M6 or 5/16-24 connection. For more information and a listing of these products, search using "53501-430".
Upchurch Scientific® Frit-In-A-Ferrule™ seal and filter simultaneously and can filter at any point in your system where ¹/₁₆ or ¹/₈" Ext.Ø tubing is used in a flat-bottom 1/4-28, M6 or 5/16-24 connection. For more information and a listing of these products, search using '53501-430'.
Most of these frits include either a PCTFE, ETFE or PEEK polymer sealing ring and many have the common 0.250"(0.64 cm) and 0.254"(0.64 cm) Ext.Øs, which allow them to be used in Upchurch Scientific® precolumn and inline filters. Choose frits with larger diameter faces and/or larger porosities for faster flow rates. Smaller diameter faces and/or smaller porosities are ideal for applications sensitive to extra flow path volume