VWR® Advanced SPP Bouffant Caps
Latex-free, White, 61 cm (24")
Supplier: VWR International
Jump to:
Product Details & Documents
Bouffant caps are manufactured from strong, breathable, spunbonded polypropylene (SPP) fabric.
- First-line barrier
- Higher basis weight for increased protection
- Non-linting
They protect against dirt, grime, and certain dry particulates in nonhazardous environments
Bouffant caps are manufactured and rigorously tested in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards. Products are validated through independent lab testing.
Caps are packaged 100 per pack, 10 packs per case.
Documents
Certificates
Buscar por número de loteTDS VWR® Advanced Protection SPP Bouffant Caps.pdf
Hoja de datos
VWR Protection Apparel
Documentación del producto
Especificaciones
- Ø:61 cm (24")
- Color:White
- Description:Latex-free
- Cat. No.:414005-001
- Particle shedding (Helmke Drum):Level I
- Weight:14.0 g/m2