Adapter, Bottom Drip, with NPT Thread, Ace Glass Incorporated
Catalog # 89052-594
Supplier: ACE GLASS, INC.
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Descripción:End-Fitting Adapter with NPT Thread, FETFE O-Ring
- Referencia:89052-594
- Para ser utilizado con:Filter Disc 89052-874
- NPT:¼"
- Grosor:#25
- Nº de proveedor:5838-78
Specifications
About this item
For use as a column end fitting that allows connection of appropriate size Ace-Thred to tubing connectors with NPT threads.
- NPT thread
- Fabricated of PTFE
- Supplied complete with disc support, screen support with retainer ring, and o-ring
Adapters are fabricated of PTFE and supplied complete with one 100 micron polyethylene disc support, one 350 micron polypropylene screen support with PTFE retainer ring, and FETFE or CAPFE o-ring. The screen with retainer ring support has a large pore size and offers less resistance to flow. However, when smaller particle size packing is used, it will be necessary to replace the screen and retainer ring with the 100 micron support.
Ordering information: Glass filter discs are also available. Contact your Avantor representative for further information. For optional filter disc remover, order 89052-532.