Pipette Tips, Sorenson BioScience
Extended Length, 50 - 1250 µl, Natural
Catalog # 89082-386
Supplier: Sorenson Bioscience
Número de proveedor #: 34750
Jump to:
Product Details & Documents
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Especificaciones
- Volume:50 - 1250 µl
- Tip Color:Natural
- Tip Style:Extended Length