Axygen® 96-Well PCR Plate, Corning
Catalog # 89005-550
Supplier: CORNING CA
Product available on GSA Advantage®
Specifications
- Nº de pocillos:96
- Faldón:Non-skirted
- Well Volume:200 µl
- Descripción:Axygen® flat top PP PCR microplate, clear, non sterile
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Reynosa
- Referencia:89005-550
- Nº de proveedor:PCR-96-FLT-C
Specifications
About this item
Axygen's plates are designed for PCR applications.
- RNase-/DNase-free
- Nonpyrogenic
- Nonsterile
- Manufactured with 99.9% pure virgin polypropylene
Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.