Corning® Advanced Polyethylene CentriStar™ Caps for Centrifuge Tubes
Catálogo # 89093-188
Proveedor: Corning
Product available on GSA Advantage®
Especificaciones
- Esterilidad:Sterile
- Volumen:15 ml
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Reynosa
- Referencia:89093-188
- Empaquetado:100/Sleeve, 500/Case
- Nº de proveedor:431471
Especificaciones
Acerca de este artículo
The CentriStar™ cap has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.
This leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions.