Corning® Centrifuge Tubes, 15 ml
Supplier: Corning
Certificates
About this item
Corning® 15 ml clear centrifuge tubes are available without cap, with the advanced Corning CentriStar™ cap or the original plug seal cap.
- Black printed graduations and a large white marking spot
- Maximum RCF is 12000×g
- Sterile, certified nonpyrogenic and DNase-/RNase-free
- 95 kPA (14 psi) pressure tested – plug seal cap only
The CentriStar™ cap has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping. This leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions.
Ordering information: Foam racks and CentriStar™ caps (catalog no. 89093-188) can be purchased separately.
Packaging: Bulk packed 25 per resealable zip lock bag.
Specifications
- Catalog No:
- 21008-673
- 21008-671
- 21008-678
- 21008-670
- 89093-186
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Related Information
Certificates
Search by Lot NumberCorning CentriStar Centrifuge Tubes Flyer Corning® CoolBox™ and CoolRack® Consumable Compatibility Guide Corning® CoolBox™ System for Consistent and Reproducible Cooling Corning® Lab Essentials Corning® LSE™ Compact Centrifuge
Product Literature
Corning Special Offer
Promotion Material
Frequently Bought Together