Corning® Aspirating Pipettes, Corning
Aspirating Pipettes, PS, Sterile, Individually Wrapped, 50/Bag, 200/Case, 5 ml, 348 mm
Supplier: Corning
Supplier #: 9099
Corning® Aspirating Pipettes, Corning
Catalog # 29442-460
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
PS pipettes, non graduated, unplugged, sterile.
- One piece PS construction
- For aseptically aspirating liquid by using vacuum suction
- Certified non pyrogenic
Packaging: Bulk (1 ml) or individually wrapped (2 and 5 ml).
Specifications
- Capacity:5 ml
- Description:Aspirating Pipettes, PS, Sterile, Individually Wrapped, 50/Bag, 200/Case
- Length:348 mm
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact
- Cat. No.:29442-460