Sample Vials with Writing Patch, Glass, Wheaton, DWK Life Sciences
Catalog # 66011-372
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Small Business Enterprise
Specifications
- Tapa incluida:Si
- Cap Material:Phenolic
- Tamaño de la tapa:13-425
- Septo:14B rubber
- Capacidad:4.00 ml
- Descripción:Sample Vials with Writing Patch
- Referencia:66011-372
- Diá. x Al:15×48 mm
- Cap Color:Black
- Nº de proveedor:225012
Specifications
About this item
Wheaton Sample Vials are made from low extractable borosilicate glass to provide superior chemical resistance.
- Autoclavable vials and caps
Vials feature a large writing patch and have attached caps to maintain cleanliness. Caps include a 14B rubber liner that is suitable for aqueous samples.
Ordering information: Vials conform to ASTM E438 Type 1, Class A and USP Type 1 standards. Packaged in Lab File® partitioned trays that provide an easy way to inventory samples or store empty vials.