Barnstead™ Smart2Pure™ Water Purification Systems, Thermo Scientific
Product Details & Documents
The Barnstead Smart2Pure water purification systems provides up to 72L/day of Type 1, ultrapure (18.2 megohm) water or Type 2 (10-15 megohm) water from tap water
All Smart2Pure systems include: quick discconnects to make changing consumables easy, and all core consumables and tubing. Smart2Pure systems are offered in three output volumes, 3L/hr, 6L/hr, and 12L/hr. The 3 and 6 LPH models come standard with an integrated reservoir. The 12 LPH model requires an external reservoir with two sizes available, 30L or 60L. The Smart2Pure systems offer highly versatile mounting options. The units can be mounted on the wall, on the bench or under the bench. For the 12 LPH model, the reservoirs may also be mounted on the wall, on the bench, or under the bench. The Smart2Pure systems are offered in a variety of configurations to meet a wide range of applications needs.
The basic Smart2Pure systems are ideal for atomic absorption, ion chromatography or other analytical procedures.
The Smart2Pure UV systems are ideal for inorganic and organic trace analysis, HPLC, or ICP-MS.
The Smart2Pure UF systems are ideal for microbiology, IVF, or monoclonal antibody applications.
The Smart2Pure UV/UF systems are ideal for molecular biology, PCR, DNA work, cell or tissue culture media.
Certifications: CSA, CE exception of 89230-328, 89230-344, 89230-346 and 89230-348.
Ordering information: Units come standard with universal power supply (90-240V, 50.60Hz) and all necessary tubing to connect to water inlet supply. The Smart2Pure 3LPH and 6LPH systems come with integrated 6L reservoir. The Smart2Pure 12LPH system requires the purchase of an external reservoir. Feed water must be tap water.
Services: Thermo Scientific provides several service options for Thermo Barnstead Water System such as Installation, Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Thermo Barnstead Water System Technical Services" in the website search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWR[email protected] for further details.
Documents
Smart2Pure Spec Table
Hoja de datos
Especificaciones
- Description:Smart2Pure UV 3LPH System (Deionization and UV), 3 L/hr output
- Cat. No.:89230-334
- W×D×H:305×400×545 mm (12×15.7×21.5")
Accessories
Información del producto
- Catalog #For Use WithAvailabilityPrice
- Filter housing includes pre filter 1 µm (Recommended for bad raw water quality)Filter housing includes pre filter 1 µm (Recommended for bad raw water quality)89230-318All Smart2Pure SystemsLog in Para disponibilidadEach (1UN) - $7,838.30
Información del producto
- Catalog #For Use WithAvailabilityPrice
- Cleaning Syringe
Información del producto
- Catalog #For Use WithAvailabilityPrice
- Final Filter, 0.2 µm for new Barnstead SystemsFinal Filter, 0.2 µm for new Barnstead Systems89230-286All GenPure xCAD SystemsLog in Para disponibilidadEach (1UN) - $9,425.00
Información del producto
- Catalog #For Use WithAvailabilityPrice
- Ultra filter for MicroPure UF, UV/UF or Smart2Pure UF, UV/UFUltra filter for MicroPure UF, UV/UF or Smart2Pure UF, UV/UF89230-458All MicroPure UF SystemsLog in Para disponibilidadEach (1UN) - $16,184.34