Orders LinkContactUsLinkComponent
Straight Plug Rubber Stoppers, Wheaton
Straight Plug Rubber Stoppers, Wheaton
Certificates
Straight Plug Rubber Stoppers, Wheaton
Straight Plug Rubber Stoppers, Wheaton
1 Options Available Below
Product Details & Documents
Wheaton straight plug stoppers provide maximum sealing for vials with a straight-wall glass finish
Stoppers are autoclavable.
Stoppers are available in a variety of sizes and rubber formulations . Straight plug stoppers are manufactured from the highest quality raw materials and are manufactured to yield exceptional dimensional stability.
Product Family Options
Información del producto
  • Fits
    Material
    Availability
    Price
  • 16171-630
    Straight plug rubber stoppers, durameter 50
    Straight plug rubber stoppers, durameter 50
    Mouth with ID×OD 15×30 mm
    Gray bromobutyl
    Discontinued
    Supplier Number: -
  • 16171-691
    Straight plug rubber stoppers, durameter 40
    Straight plug rubber stoppers, durameter 40
    Mouth with ID×OD 15×30 mm
    Natural red rubber
    Discontinued
    Supplier Number: -
  • 89168-268
    Straight plug rubber stoppers, durameter 45
    Straight plug rubber stoppers, durameter 45
    Mouth with ID×OD 13×20 mm
    Gray chlorobutyl
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $11,781.00
    $11,781.00
    Case (1,000UN)
       $11,781.00
    Supplier Number: W224100-173