Flint Glass Y-Shaped Connecting Tubes
470235-774
- Ext.Ø:5/16"
- Ø int.:1/4"
- Descripción:Flint Glass Y-Shaped Connecting Tubes, Arm Length: 1 1/2"
- Matriz:Flint glass
- Referencia:470235-774
Y-shaped connector with a 120° angle provides safer connections as the ends have constrictions to hold rubber tubing securely.
- Made from quality flint glass