XXXX Quality Corks
Supplier: MANTON INDUSTRIAL CORK SE
About this item
XXXX Quality Corks have a fine grain and are ideal for use as tight-fitting stoppers.
- Fine Grain
- Ideal for Use as Tight-Fitting Stoppers
- Common Sizes
- Regular Length
The listed sizes listed are common in school science laboratories.
The corks are regular length.
Caution: Product contains latex. May cause allergic reactions.
7 Options Available Below
Product Details & Documents
XXXX Quality Corks have a fine grain and are ideal for use as tight-fitting stoppers.
- Fine Grain
- Ideal for Use as Tight-Fitting Stoppers
- Common Sizes
- Regular Length
The listed sizes listed are common in school science laboratories.
The corks are regular length.
Caution: Product contains latex. May cause allergic reactions.
Specifications for Product Family
Specifications
- Catalog No:
- 470204-536
- 470204-424
- 470204-426
- 470204-538
- 470204-540
- 470121-846
- 470121-848
- 470121-834
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
Specifications
Product Family Options
Product Information
- Stopper No.AvailabilityPrice
- Supplier Number: 50424Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.470204-536Environmentally PreferableLow Manufacturing ImpactLow Manufacturing Impact - Supplier Number: 50524Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.470204-424Environmentally PreferableLow Manufacturing ImpactLow Manufacturing Impact - Supplier Number: 50624Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.470204-426Environmentally PreferableLow Manufacturing ImpactLow Manufacturing Impact - Supplier Number: 50724Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.470204-538Environmentally PreferableLow Manufacturing ImpactLow Manufacturing Impact - Supplier Number: 50824Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.470204-540Environmentally PreferableLow Manufacturing ImpactLow Manufacturing Impact - Supplier Number: 55511Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.470121-846Environmentally PreferableLow Manufacturing ImpactLow Manufacturing Impact - Supplier Number: 55512Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.470121-848Environmentally PreferableLow Manufacturing ImpactLow Manufacturing Impact - 18Discontinued-Supplier Number: 54520Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.470121-834Environmentally PreferableLow Manufacturing ImpactLow Manufacturing Impact