Orders LinkContactUsLinkComponent
Wire Triangles
Wire Triangles
Wire Triangles
Wire Triangles
Perfect tool for suspending a crucible.
  • Uniform clay tubes
  • Galvanized iron wire resists rusting
Product Family Options
  • Description
    Ø
  • Side length: 38 mm
    20 mm
    Paquete de 10
    Paquete de 10 - $295.59
    $295.59
    Case de 10 (10UN)
    Case de 10 (10UN) - $322.19
    $322.19
    Paquete de 10
       $295.59
    Case de 10 (10UN)
       $322.19
     : 4-2019-CS
  • Side length: 51 mm
    26 mm
    Paquete de 10
    Paquete de 10 - $343.96
    $343.96
    Case de 10 (10UN)
    Case de 10 (10UN) - $374.92
    $374.92
    Paquete de 10
       $343.96
    Case de 10 (10UN)
       $374.92
     : 4-2020-CS
  • Side length: 64 mm
    33 mm
    Paquete de 10
    Paquete de 10 - $419.19
    $419.19
    Case de 10 (10UN)
    Case de 10 (10UN) - $456.92
    $456.92
    Paquete de 10
       $419.19
    Case de 10 (10UN)
       $456.92
     : 4-2030-CS