9 V Battery Connectors
9 V battery connector, tinned wire lead terminal, 15 cm (6") leads
Catalog # 470101-278
Supplier: MOUSER ELECTRONICS SE
Número de proveedor #: 534-232
Jump to:
Product Details & Documents
Snap on battery connectors for your electronics projects.
- Alligator clips and banana plug leads offer unique connection opportunities
Power your small electronics projects with a 9 V battery. Many projects require simple tinned wire leads, but others require alligator clips or banana plugs.
Especificaciones
- Description:9 V battery connector, tinned wire lead terminal
- Length:15 cm (6") leads