Orders LinkContactUsLinkComponent
Ward's® Winogradsky Column Kit
Ward's® Winogradsky Column Kit
WARD’S Winogradsky Column Kit
Catalog # 470024-112
Supplier:   Avantor
Ward's® Winogradsky Column Kit
Catalog # 470024-112
Supplier:  Avantor
Ward's® Winogradsky Column Kit
Ward's® Winogradsky Column Kit
WARD’S Winogradsky Column Kit
Price
$433.01
Log in Para disponibilidad
KIT(1UN) Kit de 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Re-create Experiment Investigating Microbial Growth in Soil
In the late 1800s, Russian scientist Sergei N. Winogradsky invented a unique apparatus for observing microbial succession — the growth of microscopic organisms in soil. Now you can perform the same experiment, as well as a variety of others, with our Winogradsky Column Kit. Simply fill both tubes with soil samples and water, and observe the development of different types of organisms over an extended period of time. Compare their growth in soil vs. soil with nutrients added, aerobic vs. anaerobic environments, clean vs. polluted environments, and much more. You can also study different soil samples you collect, the sulfur cycle, or even protists. It comes with calcium sulfate, calcium carbonate, two columns with stoppers, and instructions with suggested experiments.
Especificaciones
  • Description:
    WARD’S Winogradsky Column Kit
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 