Orders LinkContactUsLinkComponent
Wide Range pH Test Strips
Wide Range pH Test Strips
pH Paper, wide range, 2×¼", Vial/100
Catalog # 470008-014
Supplier #:  PHP100
Wide Range pH Test Strips
Catalog # 470008-014
Wide Range pH Test Strips
Wide Range pH Test Strips
pH Paper, wide range, 2×¼", Vial/100
Price
$55.96
Log in for availability
Paquete de 100 (1UN)
Product Details & Documents
Ideal for classroom use, this general purpose pH paper provides a distinct color match for each pH unit from 1 to 14.
  • Convenient storage case
  • Easy to read color code
  • Aligned with GLOBE protocols
100 strips, measuring 2" long x 1/4" wide, are packaged in a plastic container along with a pH color matching chart.
Ordering information: Each container includes 5 books of 20 strips.
Specifications
  • Description:
    pH Paper, wide range, 2×¼", Vial/100