Weatherlink Datalogger
Data Logger, Windows, Weatherlink
470012-300
: 6510USB
Weatherlink connects your Weather Vue weather station I to an IBM/compatible or Mac.
- Create graphs, calculate average weather conditions, view weather bulletins instantly, analyze trends, and much more
- Aligned with GLOBE protocols
Weatherlink even stores data until it is transferred to your computer and can export data to Lotus 1–2–3, dBase III–compatible spreadsheet, or database software.
System Requirements–Mac: OS X, USB serial adapter; Windows: Windows 95/98/NT 4.0/2000, 5MB hard drive space, 4MB RAM, serial port.
- Description:Data Logger, Windows, Weatherlink