Orders LinkContactUsLinkComponent
Watershed Tour
Watershed Tour
Watershed Tour
Certificates
Watershed Tour
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Introduce Students to Stream and River Ecosystems
  • No liquid compounds needed for testing
  • Includes supporting instructional materials and a digital manual
  • Linked to a real-world scenario
Each virtual tour is a comprehensive curriculum designed to reinforce key concepts of water quality testing. It uses safe, nontoxic TesTabs® to test various factors of water quality. Plus, it includes a digital manual alongside lecture materials, illustrated handouts, teacher tips, test procedures, data sheets, and interactive games - making water quality education more engaging than ever.
Study a virtual river at four different points, from the headwaters to the mouth, with this classroom-based tour. Students will explore stream ecology, water quality concerns, and the impact of human activity on water health. Materials provided for 50 students.
Product Family Options
Product Information
  • No. of Lab Groups
    Availability
    Price
  • 470004-816
    Watershed Tour Lab Activity
    Watershed Tour Lab Activity
    30
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $4,037.53
    $4,037.53
    Each (1UN)
       $4,037.53
    Supplier Number: 5974
  • 470004-818
    Watershed Tour Lab Activity, Reagent Refill
    Watershed Tour Lab Activity, Reagent Refill
    30
    Discontinued
    Supplier Number: R-5419
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.