Orders LinkContactUsLinkComponent
Waterproof Field Notebooks
Waterproof Field Notebooks
Supplier:   J.L. DARLING LLC.
Waterproof Field Notebooks
Supplier:  J.L. DARLING LLC.
Certificates
Waterproof Field Notebooks
Waterproof Field Notebooks
3 Options Available Below
Product Details & Documents
An absolute must when conducting field studies.
  • Take legible notes in any conditions
  • Easy to spot if accidentally dropped
  • Aligned with GLOBE protocols
Unique all–weather paper lets you take and keep legible notes regardless of the conditions, and the bright yellow cover is easy to spot if accidentally dropped. The field version has one page for data values and on the facing page it has a grid for drawing.
Product Family Options
Información del producto
  • Width
    Length
    Availability
    Price
  • 470152-782
    Waterproof Field Notebooks, Field; 32 Sheets
    Waterproof Field Notebooks, Field; 32 Sheets
    4⅝"
    7"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $347.73
    $347.73
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $379.03
    $379.03
    Each (1UN)
       $347.73
    Paquete de 12 (1UN)
       $379.03
    Supplier Number: 353
  • 470001-928
    Waterproof Field Notebooks, Field; 78 Sheets
    Waterproof Field Notebooks, Field; 78 Sheets
    4⅝"
    7¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $836.81
    $836.81
    Log in Para disponibilidad
    Paquete de 6 (1UN)
    Paquete de 6 (1UN) - $912.13
    $912.13
    Each (1UN)
       $836.81
    Paquete de 6 (1UN)
       $912.13
    Supplier Number: 350F
  • 470152-784
    Waterproof Field Notebooks, Shirt-Pocket; 50 Sheets
    Waterproof Field Notebooks, Shirt-Pocket; 50 Sheets
    3"
    5"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $173.07
    $173.07
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $188.65
    $188.65
    Each (1UN)
       $173.07
    Paquete de 12 (1UN)
       $188.65
    Supplier Number: 135
  • Discontinued
    Supplier Number: -