Orders LinkContactUsLinkComponent
Watchmaker Forceps
Watchmaker Forceps
Watchmaker Forceps
Watchmaker Forceps
Ultra fine control.
  • Super fine point for precise work
  • Smooth surface prevents damage to delicate tissues
  • Broad handle for easy control
This is the traditional instrument for fine dissection work, with fine points and a broad handle. Choose between the standard broad handled, and economy narrow handled versions, both with very sharp, smooth tips.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
  • Description
    Material
    Length
    Tip Style
  • Watchmaker Forceps
    Stainless steel
    114 mm (4.25")
    Straight
    Each (1UN)
    Each (1UN) - $61.85
    $61.85
    Each (1UN)
       $61.85
     : 140505
  • Economy Watchmaker Forceps
    Each (1UN)
    Each (1UN) - $66.62
    $66.62
    Each (1UN)
       $66.62
     : 141400